Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. p-7184254
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111002 100 μL
PIPA5111003 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111002 Supplier Invitrogen™ Supplier No. PA5111002
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVLRDGAGKY FLWVVKFNSL NELVDYHRST SVSRNQQIFL RDIEQVPQQP T Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 100%.

GRB 2 protein contains one SH2 domain flanked on both sides by an SH3 domain. In response to EGF, GRB 2 binds, through its SH3 domains, to the guanine nucleotide exchange factor, Sos. The GRB 2-Sos complex then binds the EGF receptor. These interactions play a critical role in the activation of the p21 ras signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRB2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRB2
Gene Accession No. P62993
Gene Alias AA408164; abundant SRC homology; adapter protein GRB2; Adapter protein GRB2-A; ASH; ASH protein; EGFRBP-GRB2; epidermal growth factor receptor-binding protein GRB2; grb2; grb2.L; grb2-a; grb2-b; Grb3-3; growth factor receptor bound protein 2; growth factor receptor bound protein 2 L homeolog; growth factor receptor-bound protein 2; Growth factor receptor-bound protein 2-A; growth factor receptor-bound protein 3; HT027; hypothetical protein LOC535298; MST084; MSTP084; NCKAP2; OTTHUMP00000166097; OTTHUMP00000166098; protein Ash; SH2/SH3 adapter GRB2; SH2/SH3 adapter GRB2-A; SH2/SH3 adaptor Grb2; XELAEV_18044055mg
Gene Symbols GRB2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human GRB2. Recombinant protein control fragment (Product #RP-104615).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2885
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.