Learn More
Synthware™ Grease-Free, Two-Piece Sublimator
GREENER_CHOICESDP
Catalog No. 31502082 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
10 mL
40 mL
60 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
31-502-082 10 mL
31-502-083 40 mL
31-502-084 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-502-082 Supplier Synthware™ Supplier No. S732510
Add to Cart
Add to Cart
A grease-free, two-piece sublimation apparatus consisting of an upper water cooled condenser and lower vessel to hold the material to be sublimed.
The sublimate collects on the lower end of the condenser which has a flat bottom. Condenser is constructed using a grease-free, threaded valve. Connection of the condenser and lower vessel via an O-ring joint enables the two parts to be gently separated for maximum product recovery. Complete apparatus consists of the cold finger condenser, lower vessel, Viton™ O-ring and stainless steel pinch clamp.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Quantity 1 Ea.
Capacity (Metric) 10 mL
Product Type Water Cooled