Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ Great White™ Polyurethane Coated Gloves with Dyneema™ Fibers
Shop All Protective Industrial Products Inc. ProductsDescription
- White seamless shell, gray polyurethane coated palm and fingers
- Washable, resistant to chemicals, water and ultraviolet light
- Seamless construction
- Gray polyurethane coated grip for superior puncture and abrasion resistance li>Color coded hems for easy size identification
- Silicone-free
- Cut Level 2 protection
- Dyneema is a brand of polyethylene fibers that are 10 to 15 times stronger than steel, more durable than polyester and a strength that is 40% greater than aramid fiber
- Cuff Style: Continuous knit
Specifications
Specifications
| Coated Area | Palm, Fingers |
| Construction | Seamless Knit |
| Coating Material | Polyurethane |
| Color | Gray, White |
| Exterior Finish | No Texture |
| Hand | Specific |
| Material | Dyneema, Nylon, Spandex |
| Product Type | Dip-coated Gloves |
| Size | X-Large |
| For Use With (Application) | Construction, Electronics Assembly, Glass Cutting, Sheet Metal Handling, and General Parts Assembly |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?