Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Green CMFDA, 5-Chloromethylfluorescein Diacetate, Fluorescent Dye, 1 mg

Catalog No. 502583252
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3252 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3252 Supplier Abcam Supplier No. AB1454591MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cell-permeable green fluorescent dye for multigenerational tracking of cellular movements.

  • Molecular weight: 464.8 Da
  • Purity: >95%
  • Cell-permeable green fluorescent dye
  • Does not affect cell viability or proliferation
  • Transforms intracellularly into a cell-impermeant, fluorescent product
  • Displays fluorescence for at least 72 hours
  • Brightly fluorescent, non-toxic and stable at physiological pH
  • Allows for multigenerational tracking of cellular movements

Specifications

Detection Location In-Cell Detection
Content And Storage Store at 4°C.
Buffer 100% Green CMFDA
Shipping Condition Ambient
Detection Method Fluorescence
Label or Dye Green Fluorescent Dye
Formulation 100% Green CMFDA
Quantity 1 mg
For Use With (Application) Cell tracking
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.