Learn More
Montana Molecular Green Gi cADDis cAMP Assay - 10mL
SDP
Supplier:  Montana Molecular X0200G010
Encompass_Preferred
Measure cyclic AMP (cAMP) signaling kinetics, screen GPCRs for Gi activity, and more with the live cell, fluorescent, cADDis cAMP assay. Add the cADDis sensor to your cells and measure robust signals the next day on an automated plate reader or fluorescence imaging system. Increasing green fluorescence indicates cAMP decrease. This assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in many primary or iPSC-derived cells. The kit includes the cAMP sensor BacMam, 500mM sodium butyrate in H2O, and a positive control receptor and agonist (D2R BacMam and 2mM quinpirole in H2O). 30mL and 60mL sizes are available as well. Other Montana Molecular tools including BacMam-packaged GPCRs can easily be co-expressed with this assay.
Catalog No. NC2600264
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.