Learn More
Montana Molecular Green PIP2 Assay Kit, 10mL
Supplier: Montana Molecular D0400G010
Measure PIP2 cell signaling kinetics with this live cell fluorescent assay. Add the PIP2 sensor to your cells and measure robust signals the next day on a fluorescence imaging system. Combine with a red Montana Molecular DAG, cAMP, or R-GECO sensor for simultaneous measurements. These assays are genetically encoded and packaged in BacMam for efficient transduction in most cell types, including primary and iPSC-derived cells. Kits include positive control receptors and agonists.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.