Learn More
Eisco™ Green Safety Goggles
SCI_ED
Catalog No. S13975 Shop All Eisco Products
Change view
Click to view available options
Lens Coating:
UV-treated
Catalog No. Lens Coating
S13975 UV-treated
1 options
This item is not returnable. View return policy
Catalog No. S13975 Supplier Eisco™ Supplier No. G103
Add to Cart
Add to Cart
This item is not returnable. View return policy
General-purpose goggles featuring side vents to prevent fogging.
  • With adjustable elastic strap
  • Fit over prescription lens
  • Protect from chemical splash and dust
Specifications
Certifications/Compliance Meets ANSI Z87.1 standards.
Lens Coating UV-treated
Description Green adjustable elastic strap
Lens Tint Clear
Product Type General-purpose Safety Goggles
For Use With (Equipment) Prescription lens
For Use With (Application) Protect from chemical splash and dust
Frame Color Green
Headband Material Elastic
Ventilation Details Side ventilators
Material Polypropylene
Disposable Reusable
Lens Material Polycarbonate
Frame Material Vinyl
UV Protection UV380
Frame Type Classic