Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Green Thermoset F217 and PTFE Lined Caps PROMO

Catalog No. 02993430PM
Change view
Click to view available options
Neck Finish:
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
No. per Case:
1400/Cs.
144/Cs.
288/Cs.
340/Cs.
500/Cs.
980/Cs.
Packaging:
Bagged
Bulk
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case Packaging
02-993-430PM 70-400 144/Cs. Bagged
02-991-008PM 83-400 340/Cs. Bulk
02-991-012PM 48-400 1400/Cs. Bulk
02-991-028PM 58-400 980/Cs. Bulk
02-993-407PM 48-400 500/Cs. Bagged
02-993-408PM 45-400 500/Cs. Bagged
02-993-409PM 53-400 500/Cs. Bagged
02-993-411PM 63-400 288/Cs. Bagged
02-993-412PM 43-400 500/Cs. Bagged
02-993-425PM 83-400 144/Cs. Bagged
36-101-0373PM 58-400 500/Cs. Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 02-993-430PM Supplier Qorpak™ Supplier No. CAP00079PR

Offer wide range of chemical compatibility and high temperature tolerance

  • Resist attack from chemicals at room temperature
  • Can be used when odor and impact resilient tight seal is required
  • With low moisture transmission rate
  • Available in ribbed and smooth styles
  • Available in bagged or bulk packing

Specifications

Color Green
Material Thermoset
No. per Case 144/Cs.
No. per Pack 12/Pk.
Packaging Bagged
Liner Material F217, PTFE
Neck Finish 70-400
For Use With (Equipment) Wide mouth bottle
Closure Type Smooth
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.