Learn More
Medipak GREEN UVLI ZIP BAGS,1000/CS
Supplier: Medipak GZ92
Green UVLI™ Zip Bags provide a convenient multi-use closure, are cost-effective, and disposable for protecting light-sensitive medications during transportation and administration. Our bags exceed the United States Pharmacopoeia Standard blocking over 90% of harmful UV light between 290nm and 450nm, so you can protect with confidence.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.