Learn More
Greiner Bio-One MiniCollect™ Capillary Blood Collection System Accessories
Description
- Capillary or funnel can be inserted into MiniCollect tubes without removing the cap
- 13 × 75mm carrier tubes facilitate bar code labeling, centrifugation, and storage
- Use amber carrier tube for sheilding light-sensitive analytes
Specifications
Specifications
| Color | Lavender |
| Product Type | EDTA Tube |
| Capacity (Metric) | 80 μL |
| For Use With (Equipment) | Greiner Vacuette MiniCollect Capillary Blood Collection System |
| Tube Size | 80 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.