Learn More
Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes
Catalog No. 22030402
Change view
Click to view available options
Capacity (Metric):
0.5 mL
0.8 mL
1 mL
Additive:
K2 EDTA
K3 EDTA
Lithium Heparin
Serum
Serum Sep
Closure Color:
Gold
Green
Lavender
Red
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Additive Closure Color
22-030-402 1 mL K3 EDTA Lavender
22-030-400 1 mL Serum Red
22-030-401 0.8 mL Serum Sep Gold
22-030-405 1 mL Lithium Heparin Green
22-030-406 0.8 mL Lithium Heparin Green
22-030-403 0.5 mL K3 EDTA Lavender
22-040-200 0.5 mL K2 EDTA Lavender
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 22-030-402 Supplier Greiner Bio-One Supplier No. 450474
This item is not returnable. View return policy
Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
  • Tubes feature closure that acts as a valve, allowing appropriate capillary or funnel to be inserted without removing the cap
  • Fit into a 13 × 75mm carrier tube to facilitate bar code labeling, centrifugation, and storage
  • Tubes with gel contain an inert barrier on the bottom that moves upward to form a stable barrier between the clot/cells and serum/plasma during centrifugation and transport
  • All tubes require MiniCollect funnel or capillary collection device (sold separately)
TRUSTED_SUSTAINABILITY
Specifications
Product Type MiniCollect Capillary Blood Collection Tube
Additive K3 EDTA
Closure Type Valved Cap
Capacity (Metric) 1 mL
Closure Included Yes
Closure Color Lavender
For Use With (Application) Small volume blood collection