Learn More
Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes
Description
Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
- Tubes feature closure that acts as a valve, allowing appropriate capillary or funnel to be inserted without removing the cap
- Fit into a 13 × 75mm carrier tube to facilitate bar code labeling, centrifugation, and storage
- Tubes with gel contain an inert barrier on the bottom that moves upward to form a stable barrier between the clot/cells and serum/plasma during centrifugation and transport
- All tubes require MiniCollect funnel or capillary collection device (sold separately)
Specifications
Specifications
| Product Type | MiniCollect Capillary Blood Collection Tube |
| Additive | K3 EDTA |
| Capacity (Metric) | 0.5 mL |
| Closure Included | Yes |
| Closure Color | Lavender |
| For Use With (Application) | Small volume blood collection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.