Learn More
Greiner Bio-One Urine System Urine Kit
SDP
Choice of two draw volumes
Supplier:  Greiner Bio-One KK27402F
Catalog No. 22-040-185
Only null left
Add to Cart
Includes: Each set includes a urine culture tube with preservative (Boric acid) and urine transfer device with 10cm straw.
Description
Description
Urine Culture Tube
  • Boric acid stabilizes the microbial count in urine for up to 24 hours
  • Hermetically sealed, sterile, virtually unbreakable and space saving
Urine Transfer Device
  • Convenient sample transfer from the collection vessel
  • Tube material is polyethylene therephthalate (PET)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.