Learn More
AnaSpec GRGDS, LC - BIOTIN LABELED
Supplier:  AnaSpec AS62347
Encompass_Preferred
The GRGDS peptide contains the amino acid sequence Arg-Gly-Asp (RGD), which has been implicated as a recognition site in interactions between extracellular matrix (ECM) molecules and cell membrane receptors. RGD-containing synthetic peptides are known to inhibit attachment of endothelial cells to substrates. GRGDS peptide inhibits angiogenesis in serum-free collagen gel culture. This synthetic peptide mimics the cellular binding site of many adhesive proteins in the extracellular matrix and causes rounding and detachment of spread cells. This peptide is biotinylated through 6-aminohexanoate (LC) as a spacer.
Catalog No. NC1885935
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.