Learn More
Novus Biologicals™ Griffonia Simplicifolia Lectin I (GSL I) Isolectin B4 Native Protein
Shop All R&D Systems Products

Description
- A biologically active protein corresponding to Griffonia Simplicifolia Lectin I (GSL I) Isolectin B4
- Source: Griffonia (Bandeiraea) simplicifolia seeds
- GSL I-B4 isolectin contains only the B subunits.
- It is a useful marker for endothelial cells from nonprimates such as mouse, rat, rabbit, and goat as well as a marker for non-peptidergic unmyelinated primary afferent neurons.
- This “B”-rich lectin preferentially agglutinates blood group B cells and is specific for alpha-galactose residues.
- Fluorescein labeled GSL I-B4 has an appropriate number of fluorochromes bound to provide the optimum staining characteristics for this lectin.
- This conjugate is supplied essentially free of unconjugated fluorescein and is preserved with sodium azide.
- Excitation maximum = 495 nm; emission maximum = 515 nm. Color = green.
Specifications
Specifications
| For Use With (Application) | Bioactivity, Immunocytochemistry/Immunofluorescence |
| Molecular Weight (g/mol) | 114 kDa |
| Purification Method | Affinity purified |
| Quantity | 0.5 mg |
| Storage Requirements | Store at 4°C in the dark. |
| Conjugate | Fluorescein |
| Buffer | 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.1 mM Ca2+ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.