Learn More
Novus Biologicals™ Griffonia Simplicifolia Lectin I (GSL I) Isolectin B4 Native Protein
Shop All R&D Systems Products

Description
- A biologically active protein corresponding to Griffonia Simplicifolia Lectin I (GSL I) Isolectin B4
- Source: Griffonia (Bandeiraea) simplicifolia seeds
- GSL I-B4 isolectin contains only the B subunits.
- It is a useful marker for endothelial cells from nonprimates such as mouse, rat, rabbit, and goat as well as a marker for non-peptidergic unmyelinated primary afferent neurons.
- This “B”-rich lectin preferentially agglutinates blood group B cells and is specific for alpha-galactose residues.
Specifications
Specifications
| Concentration | Lyoph |
| For Use With (Application) | Bioactivity, Dot Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot |
| Molecular Weight (g/mol) | 114 kDa |
| Purification Method | Affinity purified |
| Quantity | 1 mg |
| Storage Requirements | Store at 4°C |
| Reconstitution | Although many buffers can be employed for reconstituting and diluting this lectin, 10 mM HEPES buffered saline, pH 8.5, 0.1 mM Ca2+ is recommended. |
| Buffer | Lyophilized from pH-adjusted water. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.