Learn More
Description
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Dot Blot, Protein Array, Immunoaffinity Purification |
| Formulation | 10 mM HEPES (pH 7.5), 0.15 M NaCl, 0.1 mM CaCl2 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 114 kDa |
| Quantity | 0.5 mg |
| Storage Requirements | Store at 4C in the dark. |
| Inhibitor Properties | For most applications, a freshly prepared working solution of 5 - 20 ug/mL in the buffer mentioned below is recommended. |
| Product Line | Novus |
| Purity or Quality Grade | Affinity purified |
| Protein | Griffonia Simplicifolia Lectin I (GSL I) Isolectin B4 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
