Learn More
Impacto™ Anti-Vibration Grip Wrap
Impacto™ 9065 Anti-Vibration tool handle grip wrap kit absorbs and dissipates shock and vibration of power tools by providing a softer, more yielding surface for optimal comfort and productivity.
Supplier:  Impacto™ 9065
Catalog No. 19-400-103
Only null left
Add to Cart
Description
Description
Impacto™ Anti-Vibration Tool Wrap Kit
  • Dampens vibration and improves tool control
  • Allows full flexibility of hand movement
  • Easy to use and customize with open or closed tool handles
  • VEP padding has the ability to stand up to, absorb and dissipate levels of repeated impact without loss of efficiency
  • VEP Dimensions 13x5x3/16 inch /33x12x0.5 cm
  • Compliance
    CE compliance; ISO 19001:2008
    Specifications
    Specifications
    Tool Grip Wrap
    13 in.
    Waffle visco-elastic polymer sheet; 2 rolls of self-adhesive silicone rubber tape
    33.02 cm
    Universal
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Promotions
    Promotions
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.