Learn More
Honeywell™ North™ Grip N™ Nitrile Coated General Purpose Polyester Work Gloves
Catalog No. 191500916
Change view
Click to view available options
Gender:
Men's
Women's
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gender
19-150-0916 Women's
19-150-0915 Men's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-150-0916 Supplier Honeywell™ North™ Supplier No. 79/1752S
This item is not returnable. View return policy
Nitrile coating on palm & back of glove provides excellent grip and a long life. Lint-free shell is made of 100% polyester and can be used in sensitive applications.
  • Nitrile N-Tread Coating improves grip, provides abrasion resistance, and durability
  • Lint-free, 100% polyester shell
  • Can be used in sensitive applications
TRUSTED_SUSTAINABILITY
Specifications
Construction String Knit
Coating Material Nitrile
Color White
Hand Ambidextrous
Material Polyester
Product Type Work Gloves
Size Universal
Style Reversible
Gender Women's
Glove Cuff Color White
Glove Cuff Style Knit
Lining Seamless Lining
Series 79/1752
Length (English) 12 in.
Length (Metric) 30.5 cm