Learn More
Honeywell™ North™ Grip N™ Uncoated Lint-Free Polyester Gloves
Catalog No. 191500917
Change view
Click to view available options
Gender:
Men's
Women's
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gender
19-150-0917 Men's
19-150-0918 Women's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-150-0917 Supplier Honeywell™ North™ Supplier No. 79/1702M
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Medium weight gloves that can be used in sensitive applications. Lint-free shell is constructed of 100% polyester.
  • Ambidextrous design gives wearer four sides for maximum economy
  • Lint-free, 100% polyester shell
  • Can be used in sensitive applications
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance 21 CFR, USDA, FDA
Construction String Knit
Coating Material Uncoated
Color White
Hand Ambidextrous
Material Polyester
Size Universal
Style Reversible
Gender Men's
Glove Cuff Style Knit
Series 79/1702
Length (English) 12 in.
Length (Metric) 30.5 cm