Learn More
Electron Microscopy Sciences Rubis Non-slip Grip Tweezers Style 5, 120 mm OAL
SDP
Supplier:  Electron Microscopy Sciences 781445
Encompass_Preferred
This special soft coating provides a non-slip grip and improves the handling of the tweezer. The soft-coating prevents hand fatigue, and protects the hand from heat and cold. Made from the finest stainless steel grade
OAL: 120 mm
Catalog No. 50-240-85
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.