Learn More
Electron Microscopy Sciences Rubis Non-slip Grip Tweezers Style 5A, 115 mm OAL
Supplier: Electron Microscopy Sciences 781445A
This special soft coating provides a non-slip grip and improves the handling of the tweezer. The soft-coating prevents hand fatigue, and protects the hand from heat and cold. Made from the finest stainless steel grade
OAL: 115 mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.