Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Grips™ Premium Goatskin Leather Drivers Gloves

Catalog No. 1146264E Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
11-462-64E Large
11-462-64C Small
11-462-64D Medium
11-462-64F X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 11-462-64E Supplier Wells Lamont™ Supplier No. Y0769L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goatskin leather is soft, pliable, and longer lasting than cow and pig leather drivers. Grips™ Premium Goatskin Leather Drivers Gloves are ideal for use in warehousing, construction, fork lift driving, and oil and gas industries.

  • Top quality goatskin with natural added lanolin for softness and extended wear
  • Goatskin has a higher natural tensile strength despite abrasion
  • Gunn cut and keystone thumb for added dexterity and flexibility
  • Bound hem

Specifications

Color Beige
Grade Premium
Hand Specific
Material Goatskin Leather
Product Type General Purpose Gloves
Size Large
Style Gunn
Glove Cuff Style Open
Hem Color Brown
Lining Unlined
Series Y0769
Thumb Style Keystone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.