Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRK6 Recombinant Rabbit Monoclonal Antibody (K01_3L85)

Catalog No. MA565866
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA565866 50 μL
MA564407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA565866 Supplier Invitrogen™ Supplier No. MA565866
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: KRKACGRQLT QNFL.

GRK6 is a member of the guanine nucleotide-binding protein coupled receptor kinase subfamily of the serine/threonine protein kinase family. GRK6 phosphorylates the activated forms of G protein-coupled receptors to initiate their deactivation. Several transcript variants encoding different isoforms have been described for GRK6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRK6
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_3L85
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene Grk6
Gene Accession No. O70293, P43250
Gene Alias activin receptor IIA; activin receptor type IIA; Activin receptor type-2A; ActrIIa; ACTR-IIA; Acvr2; Acvr2a; EC 2.7.11.16; G protein-coupled receptor kinase 6; g protein-coupled receptor kinase GRK6; Gprk6; G-protein coupled receptor kinase 6; GRK6; kinase GRK6; TactrII
Gene Symbols Grk6
Host Species Rabbit
Immunogen A synthetic peptide of human GRK6 (aa 97-110).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105540, 26385, 2870
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.