Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRLF1/p190RhoGAP Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501559572
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9572 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9572 Supplier Bethyl Laboratories, Inc Supplier No. A301736A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Dog

Represses transcription of the glucocorticoid receptor by binding to the cis-acting regulatory sequence 5′-GAGAAAAGAAACTGGAGAAACTC-3′. May participate in the regulation of retinal development and degeneration. May transduce signals from p21-ras to the nucleus, acting via the ras GTPase-activating protein (GAP). May also act as a tumor suppressor.

Specifications

Antigen GRLF1/p190RhoGAP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ARHGAP35
Gene Accession No. Q91YM2, Q9NRY4
Gene Alias ARHGAP35; glucocorticoid receptor DNA binding factor 1; Glucocorticoid receptor DNA-binding factor 1; Glucocorticoid receptor repression factor 1; GRF-1; GRF1; GRLF1; KIAA1722; MGC10745; p190-A; P190A; p190ARhoGAP; p190RhoGAP; Rho GAP p190A
Gene Symbols Arhgap35
Host Species Rabbit
Immunogen Between 1200 and 1250
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232906, 2909
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.