Learn More
Vernier™ Grommets for CO2 Gas Sensor
SDPSCI_ED
Catalog No. S16127ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Gas Sensors and BioChambers
Catalog No. For Use With (Equipment)
S16127ND Gas Sensors and BioChambers
1 options
Catalog No. S16127ND Supplier Vernier™ Supplier No. CO2GROM
Add to Cart
Add to Cart
Includes
Grommets for CO2Gas Sensor
This package of 10 grommets fit Vernier CO2 Gas Sensors and O2Gas Sensors, allowing them to be inserted into a closed system such as the Vernier BioChambers. Use them as replacement grommets for BioChambers or to create your own sample chambers.
  • This package of 10 grommets fit CO2 Gas Sensors and O2 Gas Sensors, allowing them to be inserted into a closed system such as the BioChambers.
  • Use them as replacement grommets for BioChambers or to create your own sample chambers.
Warranty and Services
This is a consumable and no warranty is included.
Specifications
Includes 10 Grommets
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) Gas Sensors and BioChambers
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov