Learn More
Echelon Biosciences Research Labs GROPI 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs P0000100UG
Encompass
Glycerophosphoinositol (GroPI) is the building block for phosphoinositide polyphosphates and is a substrate for PI-PLC. This item is non returnable
Catalog No. 50-720-543
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.