Learn More
Grainger NVENT ERICO Pointed End Ground Rod, 5/8"
Supplier:  Grainger 2KXL7
Encompass_Preferred
The NVENT ERICO Pointed End Ground Rod is a 5/8" diameter by 8' long copper-bonded steel ground rod used for grounding electrical systems. It has a pointed end for easy driving into soil and meets UL 467 requirements. The ground rod has low electrical resistance and provides a path to earth ground. It is made in the USA and can be driven into most soil types. The ground rod comes with a 5 year limited warranty from nVent.
Catalog No. NC2112646