Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Growth Hormone R (GHR) Protein, His Tag, ultra sensitivity (primary amine labeling)
Supplier:  ACROBiosystems GHRH8222200UG
Encompass_Preferred
Recombinant Protein;200UG;MABSol Biotinylated Human GHR, His Tag, ultra sensitivity (primary amine labeling) (GHR-H8222) is expressed from human HEK293 cells. It contains AA Ala 27 - Tyr 264 (Accession # P10912-1). It is the biotinylated form of Human Growth Hormone R (GHR) Protein (GHR-H5222).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.6 kDa. The protein migrates as 40-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GH, Tag Free at 2 μg/mL (100 μL/well) can bind Biotinylated Human GHR, His Tag, ultra sensitivity (primary amine labeling) (Cat. No. GHR-H8222) with a linear range of 0.5-8 ng/mL (QC tested).
Catalog No. 50-195-7540
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.