Learn More
Bachem GRP (human)
SDP
Supplier:  Bachem 4011693.1000
Encompass_Preferred
GRP (human) , H-Val-Pro-Leu-Pro-Ala-Gly-Gly-Gly-Thr-Val-Leu-Thr-Lys-Met-Tyr-Pro-Arg-Gly-Asn-His-Trp-Ala-Val-Gly-His-Leu-Met-NH2, 93755-85-2, C130H204N38O31S2, Mr 2859.42, 1mg. Gastrin Releasing Peptide, human. GRP, a bombesin-like peptide, is involved in stress-induced anorexia.
Catalog No. 50-194-6967
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.