Learn More
Thomas Scientific Pycnometer, 5 ml (+ / -0.03)
SDP
Supplier:  Thomas Scientific 8350B22
Encompass
This item has a minimum order quantity of 10 per supplier requirements
Gay-Lussac With capillary glass stopper ground to flat-sided point for take-off of overflow 10 mL and 25 mL sizes are for ASTM Standard D 369 and Barrett test H-8; AASHTO standard T-74
Catalog No. NC0444599