Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRWD1/WDR28 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501560739
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0739 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0739 Supplier Bethyl Laboratories, Inc Supplier No. A301577AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

GRWD1 is a component of the 50S and 80S preribosomal complexes and plays a role in ribosome biogenesis (Gratenstein et al., 2005 [PubMed 15885502]).

Specifications

Antigen GRWD1/WDR28
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene GRWD1
Gene Accession No. Q9BQ67
Gene Alias CDW4, GRWD, RRB1, WDR28, CUL4- and DDB1-associated WDR protein 4, WD repeat domain 28, glutamate rich WD repeat protein GRWD, glutamate-rich WD repeat-containing protein 1, regulator of ribosome biogenesis 1 homolog
Gene Symbols GRWD1
Host Species Rabbit
Immunogen Between 350 and 400
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83743
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.