Description
- Removable porous filter plate for cleaning and reuse
- Allows fine filtration without use of filter paper
Specifications
Specifications
| Capacity (Metric) | 80 mL |
| Material | Borosilicate Glass |
| Porosity | 3, Fine |