Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSDMC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116594
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116594 Supplier Invitrogen™ Supplier No. PA5116594
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa Immunogen sequence: MPSMLERISK NLVKEIGSKD LTPVKYLLSA TKLRQFVILR KKKDSRSSFW EQSDYVPVEF SLNDILEPSS SVLETVVTGP FHFSDIMIQK HKADMGVNVG.

The N-terminal moiety promotes pyroptosis. May be acting by homooligomerizing within the membrane and forming pores (PubMed:27281216). The physiological relevance of this observation is unknown (Probable). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSDMC
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 8.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene GSDMC
Gene Accession No. Q99NB5, Q9BYG8
Gene Alias gasdermin C; gasdermin C1; gasdermin-C; Gasdermin-C1; Gsdmc; Gsdmc1; melanoma-derived leucine zipper, extra-nuclear factor; melanoma-derived leucine zipper-containing extranuclear factor; MLZE; mMLZE
Gene Symbols GSDMC
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human GSDMC (NP_1136031).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56169, 83492
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.