Learn More
GSDMD Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305736AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Gasdermin D is a member of the gasdermin family. Members of this family appear to play a role in regulation of epithelial proliferation. Gasdermin D has been suggested to act as a tumor suppressor. Alternatively spliced transcript variants have been described.Specifications
GSDMD | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P57764 | |
GSDMD | |
Between 434 and 484 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
GSDMD | |
DF5L, DFNA5L, GSDMDC1, gasdermin domain containing 1, gasdermin domain-containing protein 1, gasdermin-D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79792 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.