Learn More
Cayman Chemical GSK126, 1346574-57-9, 5 mg
SDP
Supplier:  Cayman Chemical 154155
Encompass_Preferred
Molecular Formula C31H38N6O2, Purity ≥98%, Formula Weight 526.7; A selective, SAM-competitive small molecule inhibitor of EZH2 methyltransferase activity (Ki = 0.57 nM; IC50 = 9.9 nM versus that of EZH1: Ki = 89 nM; IC50 = 680 nM); inhibits global H3K27me3 levels, inhibiting the proliferation of EZH2 mutant DLBCL cell lines (IC50 = 28-61 nM) as well as the growth of EZH2 mutant DLBCL xenografts in mice receiving a daily dose of 50 mg/kg.
Catalog No. NC0761027
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.