Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 137647

Catalog No. 525750
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-575-0 50 mg
52-571-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-575-0 Supplier Tocris Bioscience™ Supplier No. 5257/50
This item is not returnable. View return policy

Potent and selective FFA4 (GPR120) agonist

GSK 137647 is a potent and selective FFA4 (GPR120) agonist (pEC50 values are 6.3, 6.2 and 6.1 at the human, mouse and rat receptor, respectively). Exhibits ≥100-fold selectivity against a panel of 61 targets including FFA1, FFA2 and FFA3. Enhances glucose-stimulated insulin secretion in MIN6 cells. Induces intracellular calcium accumulation in U2OS cells; this activity is inhibited by AH 7614 (Cat. No. 5256).

Specifications

Purity >99%
Quantity 50 mg
Product Type GSK 137647
Molecular Weight (g/mol) 305.39
Inhibitors GSK 137647

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.