Learn More
Selleck Chemical LLC GSK'872 (GSK2399872A) 5mg 1346546-69-7
SDP
Supplier:  Selleck Chemical LLC S84655MG
Encompass_Preferred
GSK'872 (GSK2399872A) is a potent and selective RIP3 kinase inhibitor which binds RIP3 kinase domain with high affinity (IC50=1.8 nM) and inhibits kinase activity with IC50 of 1.3 nM. It has minimal cross-reactivity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8004
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.