Learn More
Selleck Chemical LLC GSK J1 10mg 1373422-53-7
SDP
Supplier:  Selleck Chemical LLC S758110MG
Encompass_Preferred
GSK-J1 is a highly potent H3K27 histone demethylase inhibitor with IC50 of 28 nM and 53 nM in cell-free assays for JMJD3 (KDM6B) and UTX (KDM6A), respectively, >10-fold selectivity over other tested demethylases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4975
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.