Learn More
Medchemexpress LLC HY-12066 10mg Medchemexpress, GSK1292263 CAS:1032823-75-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1206610MG
Encompass_Preferred
Medchemexpress, HY-12066 10mg GSK1292263 CAS:1032823-75-8 GSK-1292263 is an orally available GPR119 agonist with pEC50s of 6.9 and 6.7 for human and rat GPR119, respectively. GSK-1292263 can be used for the research of type 2 diabetes mellitus (T2DM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0948
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.