Learn More
TARGETMOL CHEMICALS INC GSK180736A 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3513
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK180736A is an effective and selective GRK2 inhibitor (IC50 0.77 uM) and >100-fold selectivity over other GRKs. It is a weak inhibitor of PKA (IC50 30 uM) but highly effective against ROCK1 (IC50 100 nM). purity: 98%
Catalog No. 50-258-0233
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.