Learn More
Selleck Chemical LLC GSK1904529A 10mg 1089283-49-7 GSK 4529
SDP
Supplier:  Selleck Chemical LLC S109310MG
Encompass_Preferred
GSK1904529A (GSK 4529) is a selective inhibitor of IGF-1R and IR with IC50 of 27 nM and 25 nM in cell-free assays, >100-fold more selective for IGF-1R/InsR than Akt1/2, Aurora A/B,B-Raf, CDK2, EGFR etc. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-798-4
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.