Learn More
Medchemexpress LLC HY-15966A 100mg Medchemexpress, GSK2110183 (hydrochloride) CAS:2070009-64-0 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY15966A100MG
Encompass_Preferred
Medchemexpress,HY-15966A 100mg GSK2110183 (hydrochloride) CAS:2070009-64-0 Formula:C18H17Cl3F2N4OS GSK2110183 hydrochloride is an orally bioavailable, selective, ATP-competitive and potent pan-Akt kinase inhibitor with Kis of 0.08/2/2.6 nM for Akt1/Akt2/Akt3 respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8478
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.