Learn More
Selleck Chemical LLC GSK2193874 10mg 1336960-13-4
SDP
Supplier:  Selleck Chemical LLC S836710MG
Encompass_Preferred
GSK2193874 is an orally active, potent, and selective blocker of transient receptor potential vanilloid 4 (TRPV4) with IC50 values of 0.04 and 0.002 ?M for hTRPV4 and rTRPV4, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5991
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.