Learn More
Selleck Chemical LLC GSK2256098 5mg 1224887-10-8 GTPL7939
SDP
Supplier:  Selleck Chemical LLC S85235MG
Encompass_Preferred
GSK2256098 is a potent, selective, reversible, and ATP competitive FAK kinase inhibitor with apparent Ki of 0.4 nM. GSK2256098 inhibits cancer cell growth and induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6103
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.