Learn More
Selleck Chemical LLC GSK2334470 50mg 1227911-45-6
SDP
Supplier:  Selleck Chemical LLC S708750MG
Encompass_Preferred
GSK2334470 is a novel PDK1 inhibitor with IC50 of ~10 nM in a cell-free assay, with no activity at other close related AGC-kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1830046
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.