Learn More
Medchemexpress LLC HY-119222 5mg Medchemexpress, GSK256073 CAS:862892-90-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1192225MG
Encompass_Preferred
Medchemexpress, HY-119222 5mg GSK256073 CAS:862892-90-8 GSK256073 is a potent, selective and orally active GPR109A agonist and a long-lasting and non-flushing HCA2 full agonist with a pEC50 of 7.5 (human HCA2). GSK256073 acutely improves glucose homeostasis via inhibition of lipolysis and has the potential for the study of type 2 diabetes mellitus (T2DM)and dyslipidemia. GPR109A: G-protein coupled receptor 109A; HCA2: hydroxy-carboxylic acid receptor 2 Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0924
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.