Learn More
Medchemexpress LLC HY-19770 5mg Medchemexpress, GSK2981278 CAS:1474110-21-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY197705MG
Encompass_Preferred
Medchemexpress, HY-19770 5mg GSK2981278 CAS:1474110-21-8 GSK2981278 is a potent and selective RORγ inverse agonist. GSK2981278 inhibits activation of the il17 promoter and interferes RORγ-DNA binding. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0884
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.