Learn More
Enzo Life Sciences GSK3 substrate (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP1510001
Encompass_Preferred
This peptide sequence, RRRAAEELDSRAGpSPQL, corresponds to the sequence of the ε subunit of elF2B surrounding Ser540, a target of GSK-3 (glycogen synthase kinase 3) phosphorylation, with the addition of a phosphoserine in the 4 position. This phosphoserine allows for more efficient phosphorylation of the second serine in the peptide(Km=49 µM for GSK-3ß, 83 µM for GSK-3α). Useful in phosphocellulose GSK-3α and -ß assays. Alternative name: Glycogen synthase kinase 3 substrate. Long Term Storage: -20°C.
Catalog No. 50-201-1259
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.