Learn More
Selleck Chemical LLC GSK3326595 (EPZ015938) 5mg 1616392-22-3
SDP
Supplier:  Selleck Chemical LLC S86645MG
Encompass_Preferred
GSK3326595 (EPZ015938) is an orally active, potent and selective inhibitor of protein arginine methyltransferase 5 (PRMT5) and potently inhibits tumor growth in vitro and in vivo in animal models. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8063
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.